missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCAR3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA534425
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
A suggested positive control is Hela cell lysate. PA5-34425 can be used with blocking peptide PEP-1467.
BCAR3 Polyclonal antibody specifically detects BCAR3 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
BCAR3 | |
Polyclonal | |
Unconjugated | |
BCAR3 | |
AI131758; And34; AND-34; Bcar3; breast cancer anti-estrogen resistance 3; breast cancer antiestrogen resistance 3 protein; breast cancer anti-estrogen resistance protein 3; dJ1033H22.2 (breast cancer anti-estrogen resistance 3); KIAA0554; novel SH2-containing protein 2; NSP2; OTTHUMP00000011961; p130Cas-binding protein AND-34; SH2 domain-containing protein 3B; SH2D3B; UNQ271/PRO308 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29815, 310838, 8412 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
O75815, Q9QZK2 | |
BCAR3 | |
A 16 amino acid synthetic peptide near the amino terminus of human BCAR3 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |