Learn More
Bcl-2 with control Mouse anti-Human, Unlabeled, Clone: Bcl-2/100, BD
Mouse Monoclonal Antibody
218.28 € - 673.00 €
Spezifikation
| Antigen | Bcl-2 with control |
|---|---|
| Klon | Bcl-2/100 |
| Anwendungen | Western Blot |
| Klassifikation | Monoclonal |
| Konjugat | Unconjugated |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
15846528
|
BD Biosciences
551107 |
50 μg |
219.00 € 218.28 € / 1 Stück Sparen 0.72 € 0.33% Rabatt |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
15866528
|
BD Biosciences
551109 |
150 μg | |||||||
Beschreibung
Bcl-2 is considered to be novel among proto-oncogenes because it blocks apoptosis (programmed cell death) in many cell types. Apoptosis is an active form of cellular suicide that typically requires new RNA and protein synthesis and is associated with distinct morphological changes including cell shrinkage, cytoplasm membrane blebbing, nuclear fragmentation and DNA degradation. Because Bcl-2 blocks apoptosis it may contribute to tumorigenisis by prolonging cell survival, rather than by accelerating the rate of cell proliferation. Human Bcl-2 protein migrates at a molecular weight of ∽26 kDa by SDS-PAGE. Bcl-2/100 recognizes a 26 kDa band representing human Bcl-2. Additional minor bands at 27-31 kDa and 18-21 kDa may also be visualized. The 27-31 kDa upper band may represent a larger isoform, whereas the 18-21 kDa lower band may be an internal translation or proteolytic product, therefore, a synthetic peptide corresponding to amino acids 41-54 (GAAPAPGIFSSQPG) of human Bcl-2 was used as immunogen. This peptide sequence is not conserved between human and mouse. Bcl-2/100 does not cross-react with mouse Bcl-2. For detection of mouse Bcl-2 refer to clone 3F11 (Cat. No. 554218), polyclonal rabbit anti-rat/mouse Bcl-2 antiserum (Cat. No. 554087), and polyclonal rabbit anti-mouse Bcl-2 antiserum (Cat. No. 554279).
Spezifikation
| Bcl-2 with control | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| IgG1 κ | |
| Affinity Purified |
| Bcl-2/100 | |
| Monoclonal | |
| Mouse | |
| Human | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.