missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcl-2 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA19015
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with canine, mouse and rat based on sequence homology.
Bcl-2 Polyclonal antibody specifically detects Bcl-2 in Human samples. It is validated for Western Blot.Spezifikation
Bcl-2 | |
Polyclonal | |
Unconjugated | |
Bcl2 | |
apoptosis regulator Bcl-2; AW986256; B cell leukemia/lymphoma 2; B cell lymphoma 2 associated oncogene; B-cell CLL/lymphoma 2; B-cell leukemia/lymphoma 2; Bcl 2; BCL2; bcl-2; BCL2 apoptosis regulator; Bcl-2 protein; BCL2, apoptosis regulator; Bcl2-like protein; C430015F12Rik; D630044D05Rik; D830018M01Rik; OTTHUMP00000163680; PPP1R50; protein phosphatase 1, regulatory subunit 50 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
596 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P10415 | |
Bcl2 | |
Synthetic peptide corresponding to the N terminus amino acids AGRTGYDNREIVMKYC. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |