missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta Tubulin Loading Control Monoclonal Antibody (BT7R), DyLight™ 680, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA516308D680
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-16308-D680 has successfully been used in immunofluorescence and Western blot applications. DyLight 680 has an excitation/emission of 692/712 nm.
Beta tubulins are one of two core protein families (alpha and beta tubulins) that heterodimerize and assemble to form microtubules. Beta-III tubulin is primarily expressed in neurons and may be involved in neurogenesis, axon guidance and maintenance. Mutations in the beta tubulin gene are the cause of congenital fibrosis of the extraocular muscles type 3. Beta-III tubulin was also detected in Sertoli cells of the testis and transiently in non-neuronal embryonic tissues. Glutamate residues at the C-terminus of beta III tubulin can be glutamylated. The precise function of such modifications is unclear. Tubulin is phosphorylated on Ser-172 by CDK1 during cell cycle progression. Ser-172 phosphorylation inhibits tubulin incorporation into microtubules.Microtubules, the major cytoskeletal elements found in all eukaryotic cells, consist of Tubulin, which is a dimer of two 55 kDa subunits: alpha and Beta. Microtubules play key roles in chromosome segregation in mitosis, intracellular transport, ciliary and flagellar bending, and structural support of the cytoskeleton. Because beta-tubulin is ubiquitously expressed in all eukaryotic cells, it is frequently used as a loading control for assays involving protein detection, such as Western blotting.Spezifikation
beta Tubulin Loading Control | |
Monoclonal | |
1 mg/mL | |
PBS with proprietary stabilizer and 0.02% sodium azide | |
A2AQ07, P04350, P07437, P09207, P09244, P09652, P09653, P69897, P85108, P99024, Q13509, Q13885, Q3ZCM7, Q4QRB4, Q7TMM9, Q922F4, Q9BUF5, Q9D6F9, Q9ERD7, Q9H4B7 | |
LOC100349792, TUBB, Tubb1, Tubb2a, Tubb3, Tubb4a, Tubb5, Tubb6, TUBB8 | |
KLH conjugated peptide of the N-terminus of Beta-Tubulin. | |
50 μL | |
Primary | |
Canine, Chicken, Human, Mouse, Non-human Primate, Rabbit, Rat | |
Antibody | |
IgG2a |
Immunocytochemistry, Western Blot | |
BT7R | |
DyLight 680 | |
Tubb3 | |
2310057H16Rik; 2810484G07Rik; 3200002H15Rik; AA408537; AI325297; AI596182; B130022C14Rik; bA631M21.2; BB220206; beta 5-tubulin; beta Ib tubulin; beta tubulin; beta tubulin 1; beta-4; beta-4 tubulin; beta-4-tubulin; beta-5; beta-5 tubulin; beta5-tubulin; betaIII-tubulin; beta-tubulin; beta-tubulin class-III; Beta-tubulin class-V; beta-tubulin class-VI; Beta-tubulin isotype I; c(beta)7 tubulin; CDCBM; CDCBM1; CDCBM5; CDCBM6; CFEOM3; CFEOM3A; class I beta tubulin; Class I beta-tubulin; class IIa beta-tubulin; class III beta-tubulin; class IVa beta-tubulin; class V beta tubulin; class V beta-tubulin; CSCSC1; dJ40E16.7; dystonia 4, torsion (autosomal dominant); DYT4; FEOM3; HSA10p15 beta-tubulin 4Q; HsT1601; hypothetical protein LOC540236; hypothetical protein LOC550458; I79_000194; I79_012097; LOC100349792; LOC100761889; LOW QUALITY PROTEIN: tubulin beta-6 chain; M(beta)1; M(beta)2; M(beta)3; M(beta)4; M(beta)5; M(beta)6; M40; M40 antibody; MGC128415 protein; MGC137802 protein; neuron-spec | |
Mouse | |
Protein A | |
RUO | |
100347858, 100348290, 100349792, 100350252, 100358689, 100855815, 102154194, 10381, 10382, 203068, 22151, 22152, 22153, 22154, 246118, 29213, 29214, 307351, 347688, 396254, 396427, 421037, 431043, 474830, 480213, 485950, 498736, 545486, 679312, 67951, 7280, 768337, 81027, 84617 | |
4° C, do not freeze | |
Liquid |