missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta Tubulin Loading Control Monoclonal Antibody (BT7R), DyLight™ 650, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA516308D650
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-16308-D650 has successfully been used for immunofluorescence. DyLight 650 has an excitation/emission of 652/672nm.
beta Tubulin Loading Control Monoclonal antibody specifically detects beta Tubulin Loading Control in Chicken, Human, Mouse, Non-human Primate, Rabbit, Rat samples. It is validated for Immunocytochemistry.Spezifikation
beta Tubulin Loading Control | |
Monoclonal | |
1 mg/mL | |
PBS with proprietary stabilizer and 0.02% sodium azide | |
A2AQ07, P04350, P07437, P09207, P09244, P09652, P09653, P69897, P85108, P99024, Q13509, Q13885, Q3ZCM7, Q4QRB4, Q7TMM9, Q922F4, Q9BUF5, Q9D6F9, Q9ERD7, Q9H4B7 | |
LOC100349792, TUBB, Tubb1, Tubb2a, Tubb3, Tubb4a, Tubb5, Tubb6, TUBB8 | |
KLH conjugated peptide of the N-terminus of Beta-Tubulin. | |
50 μL | |
Primary | |
Chicken, Human, Mouse, Non-human Primate, Rabbit, Rat | |
Antibody | |
IgG2a |
Immunocytochemistry | |
BT7R | |
DyLight 650 | |
TUBB8 | |
2310057H16Rik; 2810484G07Rik; 3200002H15Rik; AA408537; AI325297; AI596182; B130022C14Rik; bA631M21.2; BB220206; beta 5-tubulin; beta Ib tubulin; beta tubulin; beta tubulin 1; beta-4; beta-4 tubulin; beta-4-tubulin; beta-5; beta-5 tubulin; beta5-tubulin; betaIII-tubulin; beta-tubulin; beta-tubulin class-III; Beta-tubulin class-V; beta-tubulin class-VI; Beta-tubulin isotype I; c(beta)7 tubulin; CDCBM; CDCBM1; CDCBM5; CDCBM6; CFEOM3; CFEOM3A; class I beta tubulin; Class I beta-tubulin; class IIa beta-tubulin; class III beta-tubulin; class IVa beta-tubulin; class V beta tubulin; class V beta-tubulin; CSCSC1; dJ40E16.7; dystonia 4, torsion (autosomal dominant); DYT4; FEOM3; HSA10p15 beta-tubulin 4Q; HsT1601; hypothetical protein LOC540236; hypothetical protein LOC550458; I79_000194; I79_012097; LOC100349792; LOC100761889; LOW QUALITY PROTEIN: tubulin beta-6 chain; M(beta)1; M(beta)2; M(beta)3; M(beta)4; M(beta)5; M(beta)6; M40; M40 antibody; MGC128415 protein; MGC137802 protein; neuron-spec | |
Mouse | |
Protein A | |
RUO | |
100347858, 100348290, 100349792, 100350252, 100358689, 10381, 10382, 203068, 22151, 22152, 22153, 22154, 246118, 29213, 29214, 307351, 347688, 396254, 396427, 421037, 431043, 498736, 545486, 679312, 67951, 7280, 768337, 81027, 84617 | |
4° C, do not freeze | |
Liquid |