missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-2 Microglobulin Monoclonal Antibody (B2M-02), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA110054
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody reacts with beta2-microglobulin associated with cell-surface MHC Class I molecules and other membrane antigens as well as with soluble beta2-microglobulin.
beta-2 Microglobulin Monoclonal antibody specifically detects beta-2 Microglobulin in Human, Porcine samples. It is validated for ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
beta-2 Microglobulin | |
Monoclonal | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P61769, Q07717 | |
B2m | |
Purified isolated human beta2-microglobulin. | |
100 μg | |
Primary | |
Human, Porcine | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
B2M-02 | |
Biotin | |
B2m | |
B2m; B2-microglobulin; b2m-W01; b2m-W03; b2m-Z01; b2m-Z02; b2m-Z03; Beta 2 microglobin; Beta 2 microglobulin; beta 2-microglobulin; beta chain of MHC class I molecules; Beta2 microglobulin; beta-2 microglobulin; beta-2 microglobulin preprotein; beta2m; beta2-m; beta-2-microglobin; beta-2-microglobulin; beta-2-microglobulin C-terminal fragment (55 AA); Beta-2-microglobulin form pI 5.3; B-F; CDABP0092; HDCMA22P; IMD43; Lactollin; Ly-m11; MHC Class I; microglobulin beta-2; Orla-B2m | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
110255236, 397033, 567 | |
4° C, do not freeze | |
Liquid |