Learn More
BMP-6 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA547695
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) BMP-7 is observed and less than 1% cross-reactivity with rhBMP-2, rhBMP-4, rhBMP-5mature, rhBMP-8A, and recombinant mouse BMP-6 is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
BMP-6 Polyclonal antibody specifically detects BMP-6 in Human samples. It is validated for Immunohistochemistry (Paraffin), Neutralization, Western Blot.Spezifikation
BMP-6 | |
Polyclonal | |
Unconjugated | |
BMP6 | |
BMP; BMP 6; BMP6; BMP-6; Bone morphogenetic protein; Bone morphogenetic protein 6; D13Wsu115e; vegetal related growth factor (TGFB-related); vegetal-related (TGFB related) cytokine; VG-1-R; VG-1-related protein; Vg1-related sequence; Vgr; Vgr1; VGR-1 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
654 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Neutralization, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P22004 | |
BMP6 | |
E. coli-derived recombinant human BMP-6 Gln382-His513 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |