Learn More
BRD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA519829
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
For Western Blot, this antibody has non-specific bands at 34 kDa and 92 kDa. This antibody is predicted to react with chicken, Xenopus laevis and zebrafish based on sequence homology.
This gene encodes a protein of unknown function. The protein contains a bromodomain, a sequence motif often found in transcriptional coactivators, and localizes to the nucleus in testis and several other cell types.Spezifikation
BRD1 | |
Polyclonal | |
Unconjugated | |
BRD1 | |
1110059H06Rik; AI316859; BR140-like protein; BRD1; BRL; Bromodomain and PHD finger-containing protein 2; bromodomain containing 1; bromodomain-containing protein 1; BRPF1; BRPF2; DKFZp686F0325; mKIAA4191; RP3-522J7.4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23774 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.9 mg/mL | |
PBS with 1% BSA and 0.02% sodium azide; pH 7.4 | |
O95696 | |
BRD1 | |
Synthetic peptide conjugated to KLH derived from within residues 150 - 250 of Human BRD1. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |