Learn More
BRD3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA530263
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: HeLa nucleus, mouse Liver. Predicted reactivity: Rat (93%), Rhesus Monkey (98%), Bovine (81%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
BRD3 Polyclonal antibody specifically detects BRD3 in Human, Mouse samples. It is validated for ChIP Assay, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
BRD3 | |
Polyclonal | |
Unconjugated | |
BRD3 | |
2410084F24Rik; AW060456; BRD3; bromodomain containing 3; bromodomain containing 3; bromodomain-containing protein 3; bromodomain containing protein 3; bromodomain-containing 3; bromodomain-containing FSH-like protein FSRG2; Bromodomain-containing protein 3; bromodomain-containing protein 3 short isoform; Fsrg2; KIAA0043; ORFX; RING3L; RING3-like protein; RINGL3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
67382, 8019 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ChIP Assay, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.54 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q15059, Q8K2F0 | |
BRD3 | |
Recombinant protein fragment corresponding to a region within amino acids 356 and 506 of Human BRD3 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |