Learn More
BRUCE Mouse, Unlabeled, Clone: 4, BD
Mouse Monoclonal Antibody
343.00 € - 610.00 €
Spezifikation
Antigen | BRUCE |
---|---|
Klon | 4 |
Konzentration | 250μg/mL |
Anwendungen | Western Blot |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15885829
|
BD Biosciences
611193 |
150 μg |
610.00 €
1 Set |
Verfügbar ab: 06-06-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15875829
|
BD Biosciences
611192 |
50 μg |
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt. Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen |
||||||
Beschreibung
Selective proteolysis is essential for the modulation of key cellular processes such as cell cycle progression. However, unlike other post-translational events, proteolysis is irreversible and therefore must occur in unidirectional cellular pathways. In eukaryotes, proteolysis is mediated primarily by the ubiquitin pathway. This pathway designates proteins for degradation by the proteasome, a multicatalytic protease complex. The ubiquitin pathway is a multistep system that tags proteins for degradation via the attachment of ubiquitin molecules to the
target. This attachment is mediated by the ubiquitin activating/conjugating enzymes E1, E2, E3, and BRUCE. BRUCE (BIR Repeat containing Ubiquitin-Conjugating Enzyme) is a novel enzyme that associates with the Golgi and the vesicular system. It contains a UBC (ubiquitin conjugating enzyme) domain, which is essential for catalysis, and a BIR (baculovirus inhibitor of apoptosis repeat) motif. BIR motifs are also found within inhibitor of apoptosis proteins (IAP) and are critical for anti-apoptotic activity. Therefore, BRUCE may function to both mediate ubiquitin-dependent proteolysis and contribute to anti-apoptotic cellular pathways.
Host Species: Mouse
Clone: 4
Isotype: IgG1
Species Reactivity: Human
Immunogen: Mouse BRUCE aa. 372-571
Formula Weight [Chemical]: 528kDa
Immunofluorescence, Western Blotting
Spezifikation
BRUCE | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified | |
BIR Repeat containing Ubiquitin-Conjugating Enzyme |
4 | |
Western Blot | |
Unconjugated | |
RUO | |
Canine, Human, Mouse, Rat | |
Mouse BRUCE aa. 372-571 | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.