missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BUBR1 Monoclonal Antibody (8G1), HRP, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA516038
Weitere Details : Netto-Gewicht : 0.10000kg
Beschreibung
This antibody should not cross-react with hBUB1.
BUBR1 Monoclonal antibody specifically detects BUBR1 in Human, Mouse samples. It is validated for Western Blot.Spezifikation
BUBR1 | |
Monoclonal | |
0.6 mg/mL | |
PBS with no preservative | |
O60566, Q9Z1S0 | |
BUB1B | |
A portion of hBUBR1 tagged at the N-terminus with pMAL (residues 1-350 of human BUBR1). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 κ |
Western Blot | |
8G1 | |
HRP | |
BUB1B | |
AU045529; BUB1 budding uninhibited by benzimidazoles 1 homolog beta; BUB1 mitotic checkpoint serine/threonine kinase B; Bub1A; Bub1b; BUB1B, mitotic checkpoint serine/threonine kinase; BUB1beta; BUBR1; budding uninhibited by benzimidazoles 1 beta; budding uninhibited by benzimidazoles 1 homolog beta; budding uninhibited by benzimidazoles 1 homolog, beta (S. cerevisiae); hBub1; hBUBR1; MAD3/BUB1-related protein kinase; MAD3L; Mitotic checkpoint kinase MAD3L; mitotic checkpoint serine/threonine-protein kinase BUB1 beta; MVA1; Protein SSK1; SSK1 | |
Mouse | |
Protein G | |
RUO | |
12236, 701 | |
4° C, store in dark | |
Liquid |