missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C10orf55 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA544486
Weitere Details : Netto-Gewicht : 0.00010kg
Descripción
Peptide sequence: DTHMELDEVH CCPGRDSGGG FIKGPMLQGL QGEGKLAPIP KPTLPSPSRL Sequence homology: Human: 100%
C10orf55 Polyclonal antibody specifically detects C10orf55 in Human samples. It is validated for Western Blot.Especificaciones
C10orf55 | |
Polyclonal | |
Unconjugated | |
C10orf55 | |
bA417O11.3; C10orf55; chromosome 10 open reading frame 55; uncharacterized protein C10orf55 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
414236 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q5SWW7 | |
C10orf55 | |
Synthetic peptide directed towards the N-terminal of human C10orf55 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |