missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1orf146 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA531672
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
C1orf146 Polyclonal antibody specifically detects C1orf146 in Human samples. It is validated for Western Blot.Spezifikation
C1orf146 | |
Polyclonal | |
Unconjugated | |
C1orf146 | |
C1orf146; chromosome 1 open reading frame 146; RP11-163M2.4; uncharacterized protein C1orf146 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
388649 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.81 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q5VVC0 | |
C1orf146 | |
Recombinant fragment corresponding to a region within amino acids 1 and 180 of Human C1orf146 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |