missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1QC Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA535084
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
C1QC Polyclonal antibody specifically detects C1QC in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
C1QC | |
Polyclonal | |
Unconjugated | |
C1QC | |
AI385742; C1qc; C1Q-C; C1qg; Ciqc; complement C1q C chain; complement C1q subcomponent subunit C; complement component 1, q subcomponent, C chain; complement component 1, q subcomponent, c polypeptide; complement component 1, q subcomponent, gamma polypeptide | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
714 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1.5 mg/mL | |
PBS with 0.09% sodium azide | |
P02747 | |
C1QC | |
KLH conjugated synthetic peptide between 93-120 amino acids from the central region of human C1QC | |
400 μL | |
Primary | |
Human | |
Antibody | |
Ig |