Learn More
C4orf14 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA531642
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: A431, Raji. Predicted reactivity: Mouse (84%), Rat (84%), Rhesus Monkey (96%), Bovine (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
C4orf14 Polyclonal antibody specifically detects C4orf14 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
C4orf14 | |
Polyclonal | |
Unconjugated | |
NOA1 | |
2610024G14Rik; AtNOS1; AU022345; C4orf14; hAtNOS1; hNOA1; mAtNOS1; mitochondrial GTPase 3 homolog; MMR_HSR1 domain containing protein RGD1359460; MNCb-4931; MTG3; nitric oxide associated 1; nitric oxide associated-1; nitric oxide synthase, mitochondrial (putative); nitric oxide-associated protein 1; Noa1; putative ortholog of Arabidopsis mitochondrial nitric oxide synthase; RGD1359460 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56412, 84273 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8NC60, Q9JJG9 | |
NOA1 | |
Recombinant fragment corresponding to a region within amino acids 276 and 510 of Human C4orf14 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |