Learn More
Complement C5b Monoclonal Antibody (aE11), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen DIA0110102
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
DIA 011-01-02 binds both membrane-bound MAC and fluid-phase SC5b-9 complexes from human, porcine, baboon and equine samples. DIA 011-01-02 has been successfully used in ELISA and IHC applications. This antibody is not recommended for Western blotting as the epitope is destroyed during the process.
Complement C5b Monoclonal antibody specifically detects Complement C5b in Equine, Human, Non-human Primate, Porcine samples. It is validated for ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).Spezifikation
Complement C5b | |
Monoclonal | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P01031, P01032 | |
C5 | |
Purified C5b-9. | |
400 μL | |
Primary | |
Equine, Human, Non-human Primate, Porcine | |
Antibody | |
IgG2a κ |
ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
aE11 | |
Unconjugated | |
C5 | |
anaphylatoxin C5a analog; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 4; C5; C5a; C5a anaphylatoxin; C5b; C5D; CHE1; complement C5; Complement C5 alpha chain; Complement C5 alpha' chain; Complement C5 beta chain; complement C5a anaphylatoxin; complement component 5; complement component C5; CPAMD4; ECLZB; FLJ17816; FLJ17822; MGC142298; prepro-C5 | |
Mouse | |
Protein A | |
RUO | |
100067655, 414437, 727 | |
4° C, store in dark | |
Liquid |