Learn More
Cathepsin E Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA547076
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This antibody detects both pro and mature mouse Cathepsin E in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 40% cross-reactivity with recombinant human Cathepsin E and less than 1% cross-reactivity with recombinant mouse Cathepsin D is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Cathepsin E Polyclonal antibody specifically detects Cathepsin E in Mouse samples. It is validated for Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot.Spezifikation
Cathepsin E | |
Polyclonal | |
Unconjugated | |
CTSE | |
A430072O03Rik; C920004C08Rik; CATE; Cathepsin E; Cathepsin E form I; Cathepsin E form II; CE; CEA; CEB; CTSE; Ctsea; EC 3.4.23.34; erythrocyte membrane aspartic proteinase; slow-moving proteinase | |
Goat | |
Antigen affinity chromatography | |
RUO | |
13034 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P70269 | |
CTSE | |
Mouse myeloma cell line NS0-derived recombinant mouse Cathepsin E Gln19-Pro397 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |