Learn More
Cathepsin L Monoclonal Antibody (193702), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA523929
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
In Western blots, less than 5% cross-reactivity with recombinant human Cathepsin L is observed and no cross-reactivity with recombinant mouse Cathepsin L is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
Cathepsin L Monoclonal antibody specifically detects Cathepsin L in Insect samples. It is validated for Immunoprecipitation, Western Blot.Spezifikation
Cathepsin L | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Mouse | |
Protein A/G | |
RUO | |
Insect | |
Antibody | |
IgG1 |
Immunoprecipitation, Western Blot | |
193702 | |
Unconjugated | |
1190035F06Rik; Cat L; cathepsin L; Cathepsin L1; Cathepsin L1 heavy chain; Cathepsin L1 light chain; CATHL; CatL; CP-2; Ctsl; CTSL1; cyclic protein 2; fs; furless; major excreted protein; MEP; nackt; nkt; p39 cysteine proteinase; Procathepsin L | |
Sf 21-derived Insect Cathepsin L | |
100 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |