Learn More
CD107a Mouse anti-Human, APC, Clone: H4A3, BD
Mouse Monoclonal Antibody
211.00 € - 715.00 €
Spezifikation
| Antigen | CD107a |
|---|---|
| Klon | H4A3 |
| Anwendungen | Flow Cytometry |
| Klassifikation | Monoclonal |
| Konjugat | APC |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
15830389
|
BD Biosciences
560664 |
50 Tests |
211.00 €
1 Stück |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
15836299
|
BD Biosciences
641581 |
100 Tests |
715.00 €
1 Stück |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
Reacts with the heavily glycosylated 110kDa lysosomal-associated membrane protein, LAMP-1. LAMP-1 is a widely expressed intracellular antigen. It is also expressed on the surface of activated platelets, PHA-activated lymphocytes and some tumor cell lines, including U937 and KG1a. LAMP-1 has been shown to be a ligand for E-selectin-mediated cell adhesion. LAMP-1 and LAMP-2 (CD107b) are carriers for poly-N-acetyllactosamines, and are able to display sialyl Le[x] termini.
Host Species: Mouse
Clone: H4A3
Isotype: IgG1, κ
Cross Reactivity: Human
Flow Cytometry
Spezifikation
| CD107a | |
| Flow Cytometry | |
| APC | |
| RUO | |
| Human Adult Adherent Peripheral Blood Cells | |
| Primary |
| H4A3 | |
| Monoclonal | |
| Mouse | |
| Human | |
| IgG1 κ | |
| Affinity Purified |
For Research Use Only.
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.