Learn More
CD120b Rat anti-Human, PE, Clone: HTNFR-M1, BD
Rat Monoclonal Antibody
Marke: BD Biosciences 552418
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
The hTNFR-M1 antibody reacts with the extracellular domain of the 75 kDa transmembrane receptor for the human cytokines, tumor necrosis factor (TNF or TNF-α) and lymphotoxin-alpha (LT-α3, aka, lymphotoxin or TNF-β). This receptor is referred to as the p75 or Type II Tumor Necrosis Factor Receptor (TNFRII) [aka, CD120b]. Human TNFRII proteins are expressed by hematopoietic cells including macrophages, neutrophils, lymphocytes, thymocytes and mast cells. TNFRII is expressed by a variety of other cell types including endothelial cells, cardiac myocytes and prostate cells. Naive B cells express very low or undetectable levels of TNFRII whereas mature erythrocytes and platelets are uniformly negative for TNFRII expression. The immunogen used to generate the hTNFR-M1 hybridoma was COS- expressed recombinant human TNFRII.
Spezifikation
CD120b | |
Monoclonal | |
PE | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
COS-expressed recombinant human TNFRII | |
100 Tests | |
Immunology | |
Human | |
IgG2b κ |
Flow Cytometry | |
HTNFR-M1 | |
TNFRSF1B; p75 TNFR; p75 TNF Receptor; TNFR80; TNF-RII; TNF-R2 | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.