Learn More
CD138 Rat anti-Mouse, PE, Clone: 281-2, BD
Rat Monoclonal Antibody
Marke: BD Biosciences 561070
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The 281-2 monoclonal antibody specifically binds to the core protein of CD138 (Syndecan-1), a cell-surface, integral membrane heparan sulfate- and chondroitin sulfate-containing proteoglycan that binds to interstitial extracellular matrix molecules. Syndecan-1 is predominantly expressed on epithelial cells, where its expression correlates with normal epithelial organization. It is also expressed on B lymphocytes at specific stages during their differentiation: precursor B cells in the bone marrow and antibody-secreting cells, including plasma cells, but not mature peripheral B cells. It is thus implicated in mediating B cell-matrix interactions. CD138 expression is also regulated during embryonic development, and the molecule shows a tissue- specific structural polymorphism resulting from different post-translational modifications. The 281-2 antibody may be used to detect the differently glycosylated forms, because it reacts with the core protein. Furthermore, the mAb detects the Syndecan-1 ectodomain which is cleaved from cell surfaces by a metalloproteinase.
Spezifikation
CD138 | |
Monoclonal | |
0.2mg/mL | |
SYND1; Syndecan-1; syn-1; Sdc1; Sstn; synstatin | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2a κ |
Flow Cytometry | |
281-2 | |
PE | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
NAMRU mouse mammary gland epithelial cell line NMuMG | |
25 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.