Learn More
CD152 Mouse anti-Human, APC, Clone: BNI3, BD
Mouse Monoclonal Antibody
265.00 € - 590.00 €
Spezifikation
Antigen | CD152 |
---|---|
Klon | BNI3 |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Konjugat | APC |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15820619
|
BD Biosciences
560938 |
25 Tests |
265.00 €
1 Stück |
Verfügbar ab: 28-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15888488
|
BD Biosciences
555855 |
100 Tests |
590.00 €
1 Stück |
Verfügbar ab: 03-06-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Reacts with the “cytolytic T lymphocyte-associated antigen”, CTLA-4. CTLA-4 is transiently expressed on activated CD28+ T cells and binds to CD80 and CD86 present on antigen presenting cells (APC) with high avidity. This interaction appears to deliver a negative regulatory signal to the T cell. There are recent reports that indicate that CTLA-4 is also expressed on B cells when cultured with activated T cells, suggesting a possible role of CTLA-4 in the regulation of B-cell response. Immobilized BNI3.1 enhances T-cell proliferation induced by CD3 and CD28. Recent studies have showed that CD152 can be expressed by regulatory T (Treg) cells. It has been found this antibody can stain the
intracellular CD152 on the Treg cells after fixation and permeabilization of cells.
Host Species: Mouse
Clone: BNI3
Isotype: IgG2a κ
Species Reactivity: Human
Flow Cytometry, Intracellular Staining
Spezifikation
CD152 | |
Flow Cytometry | |
APC | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
IgG2a κ | |
Affinity Purified | |
CTLA-4; AILIM; Cytotoxic T-lymphocyte protein 4 |
BNI3 | |
Monoclonal | |
Mouse | |
Human | |
Human CTLA4 Recombinant Protein | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.