Learn More
CD154 Mouse anti-Human, APC, Clone: TRAP1, BD
Mouse Monoclonal Antibody
267.00 € - 725.00 €
Spezifikation
Antigen | CD154 |
---|---|
Klon | TRAP1 |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Konjugat | APC |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15880629
|
BD Biosciences
560955 |
25 Tests |
267.00 €
1 Stück |
Verfügbar ab: 24-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15828378
|
BD Biosciences
555702 |
100 Tests |
725.00 €
1 Stück |
Verfügbar ab: 24-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Reacts with CD40 ligand (CD40L), a 39kDa type II membrane glycoprotein expressed on activated T cells. CD40-CD40L interaction plays a very important role in T cell-dependent B-cell proliferation, differentiation, and memory cell formation. Anti-CD40L mAb partially blocks T cell-B cell interaction affecting subsequent proliferation, differentiation, and memory cell formation. Anti-CD40L mAb partially blocks T cell-B cell interaction affecting subsequent proliferation, IL-2R expression and differentiation of B cells. In addition, blocking of CD40-CD40L interaction has been demonstrated with soluble CD40, resulting in the inhibition of immunoglobulin isotype switching. It has been reported that patients with X-linked hyper-IgM syndrome have defective expression of functional CD40L due to a defective gene that encodes CD40L. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: TRAP1
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry
Spezifikation
CD154 | |
Flow Cytometry | |
APC | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
TRAP1 | |
Monoclonal | |
Mouse | |
Baboon, Cynomolgus, Human, Rhesus Monkey | |
IgG1 κ | |
Affinity Purified | |
CD40LG; CD40 ligand; CD40L; TNFSF5; T-BAM; TRAP |
For Research Use Only.