Learn More
CD158b Mouse anti-Human, PE, Clone: CH-L, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 561723
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Reacts with a 50 - 58 kDa type I glycoprotein molecule of the killer cell inhibitory receptor (KIR) family of proteins. CD158b has also been known as GLI83, p50.2, or p58.2. CD158b is composed of two extracellular Ig-like domains. It is expressed on a subset of NK cells and it may also be present on rare T cells (either CD3/TCR αβ+, or CD3/TCR γδ+). CD158b regulates NK cell mediated cytolytic activity by interacting with HLA-C alleles (Cw 1, 3, 7 and 8).
Spezifikation
CD158b | |
Monoclonal | |
PE | |
Mouse | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
CH-L | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Human | |
IgG2b κ |
For Research Use Only.