Learn More
CD20 Mouse anti-Human, PE, Clone: 2H7, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 555623
Weitere Details : Netto-Gewicht : 0.02000kg
Description
The 2H7 monoclonal antibody specifically binds to CD20 that is encoded by the MS4A1 (Membrane-spanning 4-domains, subfamily A, member 1) gene. CD20 is a 33-37 kDa unglycosylated four-transmembrane phosphoprotein. CD20 is expressed on pre-B-cells, resting and activated B cells and follicular dendritic cells but not on plasma cells. Low level CD20 expression is observed on a small subset of normal circulating T lymphocytes. The CD20 molecule is involved in the regulation of B-cell activation.
Specifications
CD20 | |
Monoclonal | |
PE | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
2H7 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human 6.16c1.3 B cell line | |
100 Tests | |
Immunology | |
Human | |
IgG2b κ |
For Research Use Only.