Learn More
CD3 Mouse anti-Human, FITC, Clone: SP34, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 556611
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Clone SP34 reacts with human form of the 20 kDa epsilon subunit of the CD3 antigen/T-cell receptor (TCR) complex. This clone also cross-reacts with a major subset of peripheral blood lymphocytes, but not monocytes or granulocytes, of rhesus or cynomolgus and pigtail macaque monkeys. The distribution on lymphocytes is similar to that observed with normal human donor lymphocytes, with the majority of CD3-positive cells being negative when dual stained with antibodies to B or NK cell markers.
Spezifikation
CD3 | |
Monoclonal | |
FITC | |
Mouse | |
50 Tests | |
Immunology | |
Baboon, Cynomolgus, Rhesus Monkey | |
IgG3 λ |
Flow Cytometry | |
SP34 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.