Learn More
CD3 Mouse anti-Human, PE-Cy5, Clone: UCHT1, BD
Mouse Monoclonal Antibody
Marke:  BD Biosciences 555334
Weitere Details : Netto-Gewicht : 0.02000kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15898068
  • 475.38 € / 1 Stück
Verfügbar ab: 02-01-2023
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
The UCHT1 monoclonal antibody specifically binds to the human CD3ε-chain, a 20-kDa subunit of the CD3/T cell antigen receptor complex. CD3ε is expressed on 70-80% of normal human peripheral blood lymphocytes and 60-85% of thymocytes. Studies from the HLDA Workshop show that this antibody is mitogenic for CD3ε-positive cells when used in conjunction with costimulatory agents such as pokeweed mitogen or anti-CD28 antibody. CD3 plays a central role in signal transduction during antigen recognition. The UCHT1 antibody stains both surface and intracellular CD3ε unlike the other CD3 clone, HIT3a, that stains only extracellular CD3ε.
Spezifikation
Spezifikation
CD3
Monoclonal
PE-Cyanine5
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
UCHT1
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Human infant thymocytes and peripheral blood lymphocytes from a Sézary Syndrome donor
100 Tests
Immunology
Human
IgG1 κ