Learn More
CD3 Mouse anti-Human, PerCP, Clone: SP34-2, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 552851
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Clone SP34-2 is a mouse IgG1 isotype monoclonal antibody, descendant of SP34 (mouse IgG3), with the same specificity and reactivity pattern as the parent clone. It cross-reacts with a major subset of peripheral blood lymphocytes, but not monocytes or granulocytes, of baboon, and rhesus, cynomolgus, and pigtail macaque monkeys. The distribution on lymphocytes is similar to that observed with normal human donor lymphocytes with the majority of CD3-positive cells being negative when dual stained with antibodies to B or NK cells markers. SP34-2 is also capable of inducing cell proliferation on both human and non-human primate PBMC.
Spezifikation
CD3 | |
Monoclonal | |
PerCP | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
SP34-2 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Purified Human CD3ε Protein | |
50 Tests | |
Immunology | |
Baboon, Cynomolgus, Rhesus Monkey | |
IgG1 λ |
For Research Use Only.