Learn More
CD4 Mouse anti-Human, PE-CF594, Clone: RPA-T4, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 562281
Weitere Details : Netto-Gewicht : 0.00840kg
Beschreibung
The RPA-T4 monoclonal antibody specifically binds to CD4, a 59 kDa single-chain transmembrane glycoprotein that is expressed on T-helper/inducer cell populations. CD4 is also expressed on thymocytes subsets and at lower levels on monocytes and macrophages. CD4 functions as a co-receptor in MHC class II-restricted antigen-induced T cell activation and as a receptor for human immunodeficiency viruses (HIV). This antibody binds to the D1 domain (CDR1 and CDR3 epitopes) of the CD4 antigen and reacts with approximately 80% of thymocytes and 45% of peripheral blood lymphocytes. RPA-T4 is capable of blocking HIV-1, gp120, and inhibits syncytium formation.
This antibody is conjugated to BD Horizon™ PE-CF594, which has been developed exclusively by BD Biosciences as a better alternative to PE-Texas Red. PE-CF594 excites and emits at similar wavelengths to PE-Texas Red yet exhibits improved brightness and spectral characteristics. Due to PE having maximal absorption peaks at 496nm and 564nm, PE-CF594 can be excited by the blue (488-nm), green (532-nm) and yellow-green (561-nm) lasers and can be detected with the same filter set as PE-Texas Red (eg 610/20-nm filter).
Spezifikation
CD4 | |
Monoclonal | |
PE-CF594 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
RPA-T4 | |
L3T4; T-cell surface antigen T4/Leu-3; W3/25; CD4 antigen (p55) | |
Mouse | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.