Learn More
CD4 Mouse anti-Human, PE, Clone: M-T477, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 556616
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Clone M-T477 reacts with the human form of a 56 kDa transmembrane glycoprotein, CD4, present on the T-helper/inducer subset of normal human donor peripheral blood lymphocytes. This clone also cross-reacts with a subset of CD3-positive peripheral blood lymphoyctes, but not monocytes, of both rhesus and cynomolgus macaque monkeys. Cross-reactivity on both lymphocytes and monocytes (weak) of baboon is also observed. The distribution of M-T477 reactivity on lymphocytes is similar for both human and monkey, with the majority of CD4-positive lymphocytes being CD8-negative and lacking reactivity with antibodies to B- or NK-cell markers.
Spezifikation
CD4 | |
Monoclonal | |
PE | |
Mouse | |
50 Tests | |
Immunology | |
Baboon, Cynomolgus, Rhesus Monkey | |
IgG2a κ |
Flow Cytometry | |
M-T477 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.