missing translation for 'onlineSavingsMsg'
Learn More

CD45 Mouse anti-Human, APC, Clone: HI30, BD

Mouse Monoclonal Antibody

Marke:  BD Biosciences 561864

Weitere Details : Netto-Gewicht : 0.02180kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 15891409

  • 1539.00 € / 1 Stück
Verfügbar ab: 19-12-2022
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

The HI30 monoclonal antibody specifically binds to the 180, 190, 205, 220 kDa protein isoforms of CD45. CD45 is encoded by the PTPRC (Protein tyrosine phosphatase receptor type C) gene. CD45, also known as the leukocyte common antigen (LCA), is a member of the protein tyrosine phosphatase (PTP) family. It is present on all human leukocytes including lymphocytes, monocytes, granulocytes, eosinophils, and thymocytes. CD45 is absent from circulating erythrocytes, platelets, or mature erythroid cells of bone marrow and non-hemopoietic tissues.

Spezifikation

Spezifikation

CD45
Monoclonal
APC
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Human Peripheral Blood Leucocytes
500 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
HI30
PTPRC; CD45R; LCA; L-CA; Leukocyte Common Antigen; B220; T200; GP180; LY5
Mouse
Affinity Purified
RUO
Human
IgG1 κ
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only.