Learn More
CD45 Mouse anti-Human, PerCP-Cy5.5, Clone: HI30, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 564106
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The HI30 monoclonal antibody specifically binds to the 180, 190, 205, 220 kDa protein isoforms of CD45. CD45 is encoded by the PTPRC ( Protein tyrosine phosphatase receptor type C) gene. CD45, also known as the leukocyte common antigen (LCA), is a member of the protein tyrosine phosphatase (PTP) family. It is present on all human leukocytes including lymphocytes, monocytes, granulocytes, eosinophils, and thymocytes. CD45 is absent from circulating erythrocytes, platelets, or mature erythroid cells of bone marrow and non-hemopoietic tissues.
Spezifikation
CD45 | |
Monoclonal | |
PerCP-Cyanine5.5 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human Peripheral Blood Leucocytes | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
HI30 | |
PTPRC; CD45R; LCA; L-CA; Leukocyte Common Antigen; B220; T200; GP180; LY5 | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
For Research Use Only.