Learn More
CD49b Mouse anti-Human, Unlabeled, Clone: 2, BD
Mouse Monoclonal Antibody
321.00 € - 629.00 €
Spezifikation
Antigen | CD49b |
---|---|
Klon | 2 |
Konzentration | 250μg/mL |
Anwendungen | Western Blot |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15885759
|
BD Biosciences
611016 |
50 μg |
321.00 €
1 Set |
Verfügbar ab: 23-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15895759
|
BD Biosciences
611017 |
150 μg |
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt. Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen |
||||||
Beschreibung
Integrins are a family of dimeric proteins that mediate cell-to-cell and extracellular matrix adhesion. They consist of a large α chain that is non-covalently associated with a smaller β chain which is used to define integrin subfamilies. These molecules exhibit a wide range of expression throughout development and adulthood. VLA-2 (very late antigen), a member of the integrin superfamily, was identified on activated T cells, but has since been reported to be on various cell types. VLA-2 is reported to be a heterodimer of integrin α2 (CD49b) and integrin β1 (CD29) subunits. The α2 chain contains a large extracellular domain, a transmembrane domain, and a short cytoplasmic tail. VLA-2 functions as a collagen receptor on platelets and fibroblasts, as well as a collagen and laminin receptor on endothelial and epithelial cells. On activated T cells, VLA-2, like LFA-1, exhibits increased number and affinity of ligand binding. Interactions of these molecules with their extracellular matrix ligands is important for directing effector T cells to their target tissues and to provide co-stimulatory signals. Thus, VLA-2 not only plays an important role in cellular adhesion, but may function in intracellular signal transmission.
Immunofluorescence, Western Blotting
Spezifikation
CD49b | |
250μg/mL | |
Monoclonal | |
Mouse | |
Immunology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG2a | |
Affinity Purified | |
Integrin α2; VLA-2α |
2 | |
Western Blot | |
Unconjugated | |
RUO | |
Human | |
Human VLA-2α aa.42-245 | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.