Learn More
CD69 Hamster anti-Mouse, PerCP-Cy5.5, Clone: H1.2F3, BD
Hamster Monoclonal Antibody
Marke: BD Biosciences 551113
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The H1.2F3 monoclonal antibody specifically binds to CD69 (Very Early Activation antigen), an 85 kDa disulfide-linked homodimer of differentially glycosylated subunits. CD69 is a C-type lectin, most closely related to the NKR-P1 and Ly-49 NK cell-activation molecules. Its expression is rapidly induced upon activation of lymphocytes (T, B, NK, and NK-T cells), neutrophils, and macrophages. CD69 is expressed also on thymocytes that are undergoing positive selection; its role in that process is unclear. H1.2F3 mAb augments PMA-induced T-cell stimulation and IFN-γ-induced macrophage stimulation. IL-2-activated NK cells express CD69, and H1.2F3 mAb induces redirected lysis of FcR-bearing target cells by NK cells.
Spezifikation
CD69 | |
Monoclonal | |
0.2mg/mL | |
VEA; Very Early Activation Antigen; AIM; Activation Induced Molecule | |
Hamster | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
H1.2F3 | |
PerCP-Cyanine5.5 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse Dendritic Epidermal T Cell Line Y245 | |
0.1 mg | |
Immunology | |
Mouse | |
IgG1 |
For Research Use Only.