Learn More
CD8a Rat anti-Mouse, PerCP-Cy5.5, Clone: 53-6.7, BD
Rat Monoclonal Antibody
Marke: BD Biosciences 551162
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The 53-6.7 antibody has been reported to react with the 38kDa α and 34kDa α′ chains of the CD8 differentiation antigen (Ly-2 or Lyt-2) of all mouse strains tested. The CD8 α and α′ chains (CD8a) form heterodimers.
Host Species: Rat
Clone: 53-6.7
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: Mouse thymus / spleen Cells
Flow Cytometry
Spezifikation
CD8a | |
Monoclonal | |
0.2mg/mL | |
Cd8a; CD8 alpha chain; Ly-2; Lyt2; Lyt-2; Ly-35; Ly-B | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
53-6.7 | |
PerCP-Cyanine5.5 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse thymus / spleen Cells | |
0.1 mg | |
Immunology | |
Mouse | |
IgG2a κ |
For Research Use Only.