Learn More
C1S Monoclonal Antibody (9), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen ABS0020902
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
ABS 002-09-02 is specific for C1s proenzyme and activated free C1s. ABS 002-09-02 has been successfully used in ELISA procedures. ABS 002-09-02 reacts in ELISA when tested with activated C1s enzyme directly coated onto the microtiter well or in sandwich ELISA in combination with a polyclonal antibody in coat. Epitope of ABS 002-09-02 differs from that of ABS 002-49-02. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL
C1S Monoclonal antibody specifically detects C1S in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin).Spezifikation
C1S | |
Monoclonal | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P09871 | |
C1S | |
C1s proenzyme purified from human plasma. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Immunohistochemistry (Paraffin) | |
9 | |
Unconjugated | |
C1S | |
AA959438; AI255193; AI327365; basic proline-rich peptide IB-1; C1 esterase; C1s; C1s1; C1sa; complement C1s; complement C1s subcomponent; Complement C1s subcomponent heavy chain; Complement C1s subcomponent light chain; Complement C1s-A subcomponent; Complement C1s-A subcomponent heavy chain; Complement C1s-A subcomponent light chain; Complement component 1 subcomponent s; complement component 1 subcomponent s-A; complement component 1, s subcomponent; complement component 1, s subcomponent 1; complement component C1SA; r-gsp; serine protease | |
Mouse | |
Protein A/G | |
RUO | |
716 | |
4° C, store in dark | |
Liquid |