missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OPRD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA124532
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Peptide sequence identity/similarity to mouse:81/87%. Rat 75/75%.
OPRD1 Polyclonal antibody specifically detects OPRD1 in Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
OPRD1 | |
Polyclonal | |
Unconjugated | |
OPRD1 | |
delta 1 opioid receptor; delta opioid receptor 1; delta-type opioid receptor; DOR; D-OR-1; DOR-1; K56; mDOR; MSL-2; Nbor; Opioid receptor; opioid receptor A; opioid receptor delta 1; opioid receptor, delta 1; OPRD; Oprd1; Ror-a | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4985 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P41143 | |
OPRD1 | |
Synthetic 16 amino acid peptide from C-terminus of human delta Opioid receptor | |
25 μg | |
Primary | |
Human | |
Antibody | |
IgG |