Learn More
eIF-4E Mouse, Unlabeled, Clone: 87, BD
Mouse Monoclonal Antibody
321.00 € - 665.00 €
Spezifikation
Antigen | eIF-4E |
---|---|
Klon | 87 |
Konzentration | 250μg/mL |
Anwendungen | Western Blot |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15885369
|
BD Biosciences
610269 |
50 μg |
321.00 €
1 Set |
Verfügbar ab: 17-06-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15895369
|
BD Biosciences
610270 |
150 μg |
665.00 €
1 Set |
Verfügbar ab: 05-06-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
The eukaryotic translation initiation factor 4E (eIF-4E) is a 25kDa phosphoprotein that specifically binds to the 7-methylguanosine-containing cap of mRNA. eIF-4E is the rate-limiting component for the initiation of cap-dependent translation by the eIF-4E translation initiation complex. This complex promotes the unwinding of secondary structure at the 5′ untranslated region of mRNA, which is necessary to expose and locate the AUG-initiation codon. Phosphorylation of eIF-4E on Ser-209 occurs after serum treatment in CHO cells, and may regulate its function. Overexpression of eIF-4E can lead to increased cell proliferation, transformation, and tumorigenesis in nude mice. The overexpression of a Ala-53 variant of eIF-4E cannot evoke these changes, suggesting that Ser-53 on eIF-4E participates in the transfer of mRNA to the 48S initiation complexes. In cooperation with nuclear oncogenes such as c-myc or E1A, eIF-4E transforms primary cells. Other studies have demonstrated that overexpression of eIF-4E causes activation of Ras and leads to a transformed phenotype. Subsequent overexpression of GAP then causes reversion of this phenotype. The mechanism by which eIF-4E plays a role in transformation is not clear, but it is postulated that high levels of eIF-4E may lead to the translation of mRNAs that are normally translationally repressed.
Host Species: Mouse
Clone: 87
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Rabbit eIF-4E aa. 1-217
Immunofluorescence, Immunoprecipitation
Spezifikation
eIF-4E | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified |
87 | |
Western Blot | |
Unconjugated | |
RUO | |
Canine, Chicken, Frog, Human, Mouse, Rat | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.