Learn More
eIF3k Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA527593
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T. Predicted reactivity: Mouse (97%), Xenopus laevis (85%), Rhesus Monkey (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
eIF3k Polyclonal antibody specifically detects eIF3k in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
eIF3k | |
Polyclonal | |
Unconjugated | |
EIF3K | |
1200009C21Rik; ARG134; eIF-3 p25; eIF-3 p28; eIF3k; EIF3-p28; Eif3s12; eukaryotic translation initiation factor 3 subunit 12; Eukaryotic translation initiation factor 3 subunit K; eukaryotic translation initiation factor 3, subunit 12; eukaryotic translation initiation factor 3, subunit K; HSPC029; M9; MSTP001; muscle specific; muscle-specific gene M9 protein; PLAC24; PLAC-24; PRO1474; PTD001 | |
Rabbit | |
Protein A | |
RUO | |
27335, 73830 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
10 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9DBZ5, Q9UBQ5 | |
EIF3K | |
Recombinant fragment corresponding to a region within amino acids 41 and 218 of Human EIF3K | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |