Learn More
eIF4G3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA531101
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, A431, HeLa, HepG2, mouse testis. Predicted reactivity: Human (99%), Mouse (96%), Rat (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is thought to be part of the eIF4F protein complex, which is involved in mRNA cap recognition and transport of mRNAs to the ribosome. Interestingly, a microRNA (miR-520c-3p) has been found that negatively regulates synthesis of the encoded protein, and this leads to a global decrease in protein translation and cell proliferation. Therefore, this protein is a key component of the anti-tumor activity of miR-520c-3p.Spezifikation
eIF4G3 | |
Polyclonal | |
Unconjugated | |
EIF4G3 | |
1500002J22Rik; 4833436O05; 4930523M17Rik; eIF4G 3; eIF-4G 3; Eif4g3; eIF-4-gamma 3; eIF-4-gamma II; eIF4GII; eukaryotic translation initiation factor 4 gamma 3; eukaryotic translation initiation factor 4 gamma, 3; G1-419-52; repro8; RP11-190H11.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
230861, 298573, 8672 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.26 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
O43432, Q80XI3 | |
EIF4G3 | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 203 of Human EIF4G3 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |