missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EML4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA517334
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
It is not recommended to aliquot this antibody.
EML4 Polyclonal antibody specifically detects EML4 in Human samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western Blot.Spezifikation
EML4 | |
Polyclonal | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
Q9HC35 | |
EML4 | |
Synthetic peptide corresponding to residues surrounding Arg91 of human EML4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
EML4 | |
4930443C24Rik; AI644019; C2orf2; echinoderm microtubule associated protein like 4; echinoderm microtubule-associated protein-like 4; ELP120; EMAP-4; EMAPL4; EML4; OTTHUMP00000201447; OTTHUMP00000201455; Restrictedly overexpressed proliferation-associated protein; ropp 120; ROPP120 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27436 | |
-20°C | |
Liquid |