missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EMR1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA533503
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human (100%) Gorilla (89%).
EMR1 has a domain resembling seven transmembrane G protein-coupled hormone receptors (7TM receptors) at its C-terminus. The N-terminus of the encoded protein has six EGF-like modules, separated from the transmembrane segments by a serine/threonine-rich domain, a feature reminiscent of mucin-like, single-span, integral membrane glycoproteins with adhesive properties. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene. EMR1 is the human ortholog of murine F4/80 but unlike F4/80, the expression of EMR1 in humans is restricted to eosinophils and is not expressed on macrophage populations.Spezifikation
EMR1 | |
Polyclonal | |
Unconjugated | |
ADGRE1 | |
ADGRE1; Adhesion G protein-coupled receptor E1; Adhesion G protein-coupled receptor E2; cell surface glycoprotein EMR1; cell surface glycoprotein F4/80; DD7A5-7; egf-like module containing, mucin-like, hormone receptor-like 1; EGF-like module containing, mucin-like, hormone receptor-like sequence 1; EGF-like module receptor 1; EGF-like module-containing mucin-like hormone receptor-like 1; EGF-TM7; Emr1; EMR1 hormone receptor; F4/80; Gpf480; Ly71; lymphocyte antigen 71; TM7LN3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2015 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q14246 | |
ADGRE1 | |
Synthetic 19 amino acid peptide from N-terminal extracellular domain of human EMR1. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |