missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EMR3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA533508
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human (100%) Gibbon (94%) Gorilla (89%) Marmoset (83%).
EMR3 Polyclonal antibody specifically detects EMR3 in Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
EMR3 | |
Polyclonal | |
Unconjugated | |
ADGRE3 | |
ADGRE3; Adhesion G protein-coupled receptor E3; egf-like module containing, mucin-like, hormone receptor-like 3; EGF-like module receptor 3; EGF-like module-containing mucin-like hormone receptor-like 3; egf-like module-containing mucin-like receptor 3; EMR3; UNQ683/PRO1562 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84658 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q9BY15 | |
ADGRE3 | |
Synthetic 18 amino acid peptide from N-terminal extracellular domain of human EMR3. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |