missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENDOG Monoclonal Antibody (7G1G10), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA515757
Weitere Details : Netto-Gewicht : 0.10000kg
Beschreibung
A suggested positive control is HepG2 cell lysate.
The protein encoded by this gene is a nuclear encoded endonuclease that is localized in the mitochondrion. The encoded protein is widely distributed among animals and cleaves DNA at GC tracts. This protein is capable of generating the RNA primers required by DNA polymerase gamma to initiate replication of mitochondrial DNA.Spezifikation
ENDOG | |
Monoclonal | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q14249 | |
ENDOG | |
Recombinant protein corresponding to amino acids 51 - 140 of human EndoG. | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG1 |
Western Blot | |
7G1G10 | |
Unconjugated | |
ENDOG | |
endo G; Endog; endonuclease G; endonuclease G, mitochondrial; mitochondrial endonuclease G | |
Mouse | |
Protein A | |
RUO | |
2021, 362100 | |
-20°C | |
Liquid |