Learn More
RECA-1 Monoclonal Antibody (RECA-1), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA140240
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA1-40240 detects Endothelial cell antigen from rat samples. MA1-40240 has been successfully used in immunohistochemistry (frozen), immunohistochemistry (paraffin), flow cytometry and immunofluorescence applications. The MA1-40240 immunogen is cell surface antigen. Positive control: Glomerular endothelial cells, RHEC cell line; Negative control: Vascular smooth muscle cells, fibroblasts
RECA-1 Monoclonal antibody specifically detects RECA-1 in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).Spezifikation
RECA-1 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.1% BSA and 0.02% sodium azide | |
Mouse | |
Protein G | |
RUO | |
Rat | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
RECA-1 | |
Unconjugated | |
Reca1 | |
Cell surface antigen. | |
100 μg | |
Primary | |
4° C | |
Liquid |