missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endothelin B Receptor Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA527173
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: Mouse brain, Rat brain. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Endothelin B Receptor Polyclonal antibody specifically detects Endothelin B Receptor in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Endothelin B Receptor | |
Polyclonal | |
Unconjugated | |
EDNRB | |
ABCDS; AU022549; Ednra; Ednrb; endothelin B receptor; endothelin receptor non-selective type; endothelin receptor subtype B1; endothelin receptor type B; endothelin-B receptor; ETb; ET-B; Etb receptor; Et-b receptor; ETB1; ETBR; ET-BR; Et-b-r; Etb-svr; ETRB; ET-RB; ETR-b; HSCR; HSCR2; piebald; RP11-318G21.1; Sox10m1; WS4A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
13618, 1910, 50672 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7 | |
P21451, P24530, P48302 | |
EDNRB | |
Synthetic peptide corresponding to a region within amino acids 8 and 111 of Endothelin B Receptor | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |