missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENTPD3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA521567
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: Molt-4, Raji. Predicted reactivity: Rhesus Monkey (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ENTPD3 Polyclonal antibody specifically detects ENTPD3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
ENTPD3 | |
Polyclonal | |
Unconjugated | |
ENTPD3 | |
CD39 antigen-like 3; CD39L3; CD39-like 3; ecto-apyrase 3; Ecto-ATP diphosphohydrolase 3; ecto-ATPase 3; ecto-ATPDase 3; Ectonucleoside triphosphate diphosphohydrolase 3; ENTPD3; HB6; NTPDase 3; NTPDase3; NTPDase-3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
956 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.41 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
O75355 | |
ENTPD3 | |
Recombinant fragment corresponding to a region within amino acids 200 and 459 of ENTPD3 (Uniprot ID#O75355) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |