missing translation for 'onlineSavingsMsg'
Learn More

Eps15 Mouse, Unlabeled, Clone: 17, BD

Mouse Monoclonal Antibody

Marke:  BD Biosciences 610806

Weitere Details : Netto-Gewicht : 0.01000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 15885649

  • 308.94 € / 1 Set
Verfügbar ab: 19-12-2022
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

The eps15 (EGF receptor pathway substrate clone number 15) gene was isolated using an expression cloning method designed for direct isolation of cDNAs encoding substrates of tyrosine kinases. The eps15 gene encodes a 145kDa protein that is tyrosine phosphorylated following EGF receptor activation and this receptor directly phosphorylates purified Eps15 in vitro. Phosphorylation of Eps15 is much more efficient following EGF receptor activation versus erbB2 kinase activation, suggesting the protein is predominantly a part of an EGF receptor signaling pathway. Overexpression of Eps15 causes transformation of NIH 3T3 cells, implying Eps15 may be involved in the control of cell proliferation. Eps15 binds to the SH3 domain of Crk through its proline-rich domain at the C-terminal region. Adaptin α binds to the C-terminus of Eps15 at three different sites ranging from residues 650 to 730. Eps15, Adaptin α and EGF receptor are co-localized at the clathrin-coated pits after receptor activation. Therefore, Eps15 may recruit EGF receptors to the clathrin-coated pits for internalization of the activated receptor.

Immunofluorescence, Immunoprecipitation, Western Blotting

Spezifikation

Spezifikation

Eps15
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Human Eps15 aa. 694-888
50 μg
Cell Biology
Rat, Mouse, Human
IgG2a
Western Blot
17
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only.