Learn More
FXR Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA540755
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: AECLLTEIQC KSKRLRKNVK QHADQTVNED SEGRDLRQVT STTKSCREKT Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%
FXR Polyclonal antibody specifically detects FXR in Human, Mouse samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
FXR | |
Polyclonal | |
Unconjugated | |
NR1H4 | |
AI957360; BAR; bile acid receptor; farnesoid X activated receptor; farnesoid X nuclear receptor; farnesoid X-activated receptor; Farnesol receptor HRR-1; FXR; HRR1; HRR-1; hypothetical protein LOC540528; MGC163445; NR1H4; Nuclear receptor subfamily 1 group H member 4; nuclear receptor subfamily 1, group H, member 4; retinoid X receptor-interacting protein 14; RIP14; RXR-interacting protein 14; Rxrip14 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
20186, 9971 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q60641, Q96RI1 | |
NR1H4 | |
Synthetic peptide directed towards the middle region of human NR1H4 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |