Learn More
GC Mouse anti-Human, Clone: 19E8, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen HYB2490502
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
HYB 249-05-02 detects Gc-Globulin in human samples. HYB 249-05-02 has been successfully used in ELISA techniques. HYB 249-05-02 reacts strongly with Gc-globulin when tested in sandwich ELISA in combination with a polyclonal antibody against Gc-globulin. The HYB 249-05-02 immunogen is Gc-Globulin found in human plasma cells. Epitope differs from that of HYB 249-01-02, HYB 249-02-02 and HYB 249-10-02. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL
GC (vitamin D-binding protein) is involved in vitamin D transport/storage, scavenging of extracellular G-actin, enhancement of the chemotactic activity of C5 alpha for neutrophils in inflammation and macrophage activation.Spezifikation
GC | |
Monoclonal | |
0.93 mg/mL | |
GC | |
Vitamin D-binding protein, DBP, Group-specific component, Gc-globulin, VDB | |
Mouse | |
Protein A | |
RUO | |
2638 | |
4° C, store in dark | |
Liquid |
ELISA | |
19E8 | |
Unconjugated | |
P02774 | |
GC | |
Gc-globulin isolated from human plasma. | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |